missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Whatman™ Qualitative Filter Papers, Wet Strengthened Grades
Description
- Fast filter paper
- Aflatoxin determination in animal feed
- Mercury determination in sludge
Specifications
Specifications
| For Use With (Application) | Aflatoxin determination in animal fee. Mercury determination in sludge. |
| Grade | 588 |
| Diameter (Metric) | 240 mm |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?