missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ Pyramid Folded Filter Papers

Easy to use, saves valuable time when undertaking repetitive or multiple analyses

Brand:  Cytiva 9892-128

Additional Details : Weight : 1.05000kg

 View more versions of this product

Product Code. 16072042

  • £297.00 / Pack of 1000
Estimated Shipment: 11-01-2024
to see stock.
Add to basket

Description

Description

  • Easily fits into conical funnels
  • Saves valuable time when undertaking repetitive or multiple analyses
  • Provides the same high quality and consistency that is expected from all Whatman™ Filter Papers
Specifications

Specifications

95 g/m2
Folded
Conical Funnels, Repetative or Multiple Anaylsis
1000/Pk.
Filter Paper
Ash Content: 0.007%
Cellulose
Grade 40
210 μm
125 mm
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers