missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ Puradisc™ 25mm Polypropylene Syringe Filters

Whatman™ Puradisc™ 25mm Polypropylene Syringe Filters may be used for biological sample and HPLC aqueous sample preparation.

Brand:  Cytiva 6786-2502

129.01 GBP valid until 2024-05-06
Use promo code "21711" to get your promotional price.



Code : 75

Additional Details : Weight : 0.17000kg

 View more versions of this product

Product Code. 11304694

  • £144.00 / Pack of 50
Estimated Shipment: 21-05-2024
to see stock.
Add to basket

Description

Description

Whatman™ Puradisc™ 25mm Polypropylene Syringe Filters may be used for biological sample and HPLC aqueous sample preparation.

  • Diameter: 25mm
  • Sample volume up to 100mL
  • Low hold-up volumes for maximum sample recovery
  • Optional glass microfiber
  • Autoclave at 121ºC
Applications:
  • HPLC aqueous sample preparation
  • Biological sample preparation
  • Buffer solutions
  • Salt solutions
  • Tissue culture media
  • Irrigation solutions
  • Sterile isolation
Specifications

Specifications

Puradisc 25 Polypropylene Syringe Filter, 0.2 μm (50 pcs)
0.2 μm
4.2 cm2
Female luer lock
Male luer
50 Pieces
Up to 100 mL
25 mm
Polypropylene
Non-sterile
Pigment Free Polypropylene (PP)
50
5.2 bar (75 psi)
Syringe Filter
22.9 x 28.4 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers