missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ Protran™ Nitrocellulose Blotting Membranes: 0.45μm BA85 Protran Circles
Description
- Universal membrane suitable for Western, Southern, and Northern blotting
- Hydrophilic, so no prewetting with methanol is required
- High binding capacity and very low background for superior signal-to-noise ratios without stringent washing conditions
- Compatible with all common detection methods, including radioactive, colorimetric, fluorescent, and chemiluminescent
- Provide excellent shelf-life for proteins—proteins maintain molecular recognition activity for up to five years on Protran membranes
- Three pore sizes available—0.45μm is ideal for larger molecular weight samples, 0.2μm ensures binding of samples below 20kDa, 0.1μm is used for molecules below 7kDa
Specifications
Specifications
| For Use With (Application) | Suitable for radioactive, colorimetric, fluorescent, and chemiluminescent detection |
| Binding Capacity | High |
| Diameter (Metric) | 82 cm |
| Quantity | 50/Pk. |
| Format | Disc |
| Material | Nitrocellulose |
| Pore Size | 0.45 μm |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?