missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ MD050 and MD142 Series Pressure Filtration Manifolds with Infusion Cylinders

Product Code. 11395004
Change view
Click to view available options
Model:
MD050/4
MD142/5/3
MD142/7/3
Material:
PTFE
Stainless Steel
Volume (Metric):
200 mL
1500 mL
2200 mL
Includes:
PTFE O-Ring, 1500 mL Infusion Cylinder for 142 mm Membrane
Silicone O-Ring/PTFE O-Ring, 200 mL Infusion Cylinder for 50 mm Membrane, Rapid Seal Connector
Silicone O-Ring/PTFE O-Ring, 2200 mL Infusion Cylinder for 142 mm Membrane
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model Material Volume (Metric) Includes unitSize
11395004 MD142/7/3 PTFE 1500 mL PTFE O-Ring, 1500 mL Infusion Cylinder for 142 mm Membrane Each
11335004 MD050/4 Stainless Steel 200 mL Silicone O-Ring/PTFE O-Ring, 200 mL Infusion Cylinder for 50 mm Membrane, Rapid Seal Connector Each
11365004 MD142/5/3 Stainless Steel 2200 mL Silicone O-Ring/PTFE O-Ring, 2200 mL Infusion Cylinder for 142 mm Membrane Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11395004 Supplier Cytiva Supplier No. 10451710

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Cytiva Pressure Filtration Manifolds come in PTFE or stainless steel. Equipped with infusion cylinders, they are suitable for batch filtration from 20 mL, filtration of liquids/gases, ‘sterile’ filtration of serum, or clear filtration of viscous media using membranes, paper, or glass fiber discs.

  • Product Type: Pressure Filtration Manifold
  • Includes: Infusion Cylinder
  • Temperature Resistance Maximum: 200°C
TRUSTED_SUSTAINABILITY

Specifications

Disposable Reusable
Material PTFE
Sterility Non-sterile
For Use With (Application) Pressure Filtration
Includes PTFE O-Ring, 1500 mL Infusion Cylinder for 142 mm Membrane
Max. Pressure 3.5 bar (PTFE O-Ring)
Model MD142/7/3
Temperature (English) Max. 392°F
Temperature (Metric) Max 200°C
Type Pressure Filtration Manifold
Volume (Metric) 1500 mL
Width (English) 7.9 in.
Width (Metric) 200 mm
Height (English) 18.5 in.
Height (Metric) 470 mm
Diameter (Metric) Filter 134 mm (Pre-filter), 142 mm (Filter)
Diameter (Metric) Inside 142 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.