missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ Polydisc Inline Filters

Versatile, cost-effective filters designed for larger volume sample filtration

Brand:  Cytiva 6720-5002

186.79 GBP valid until 2025-03-30
Use promo code "24134" to get your promotional price.


 View more versions of this product

Product Code. 11314634

  • £226.00 / Pack of 10

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Versatile, cost-effective filters designed for larger volume sample filtration

  • Filter up to 1L with one device
  • Can be used in conjunction with a syringe or connected inline (6-10mm I.D. hose) via stepped hose barbs
  • High-purity polypropylene housing maintains sample purity
  • 50mm filter devices
  • Autoclavable
  • Lightweight (11.5g)
Polydisc AS (Aqueous Solution)
  • For tissue culture media, reagent preparation, particle counting solutions, pharmaceutical preparations High-throughput polyethersulfone membrane (low protein binding and no surfactants)
  • Glass microfiber prefilter extends membrane life and effectively filters heavily contaminated samples
  • Sterility cap on outlet (optional)
  • No EtO residuals Barbed hose connections fit multiple tubing sizes Integrity testable (bubble point method) Sealed in its own “medical grade” clear blister pack and secured in a protective shelf pack
Polydisc TF
  • Use in vents, inline, exhausts for growth environments, clean or sterile gases, solvents, reagents, drying gases, photoresists Hydrophobic PTFE membrane ideal for chemically aggressive solutions, reagents and organic solvents
  • Chemical-resistant housing Integrity testable (bubble point or water breakthrough pressure in situ methods) Biosafe

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Autoclavable
PTFE
16 cm2
Polypropylene
Pk. of 10
50 mm
Polydisc TF Filter, polypropylene, 0.2 μm (10 pcs)
0.2 μm
Larger volume sample filtration
Tubing Nozzle 6 to 10 mm i.d. hose
Polydisc TF
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.