missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ PVP-Free Polycarbonate Cell Culture Membrane
Combines PVP-free polycarbonate with sharp, uniform cylindrical pores. The Whatman™ PVP-Free Polycarbonate Cell Culture Membrane achieves a delicate balance between cell adhesion and migration for effective chemotaxis.
Brand: Cytiva 155845
Code : 69
Additional Details : Weight : 0.06000kg
Description
- Enhanced cellular adhesion in biological applications such as neutrophil chemotaxis
- Cell culture filter suitable for biological applications
- Thin and uniform cylindrical pores for rapid cell migration
- Offered without the standard PVP wetting agent for increased cellular adhesion
- Reduces incubation time and the need to sterilize
Specifications
Polycarbonate | |
High | |
80 x 25mm |
5 μm | |
Epifluorescence microscopy, environmental analysis, cell biology |