missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ pH Indicators

Product Code. 11364974
Change view
Click to view available options
pH Range:
0 to 14
1 to 11
2 to 9
Quantity:
1 Roll
100 Pack
200 Pack
Unit Size:
Each
Pack of 100
Pack of 200
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. pH Range Quantity unitSize
11364974 2 to 9 100 Pack Pack of 100
11354974 0 to 14 100 Pack Pack of 100
11919347 1 to 11 1 Roll Each
11375254 0 to 14 200 Pack Pack of 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11364974 Supplier Cytiva Supplier No. 10362010
Add to basket
Add to basket
This item is not returnable. View return policy

Provides rapid determination of pH values. Whatman™ pH Indicators are impregnated with a mixture of several indicators that change color when they come in contact with a particular sample solution.

Determine pH using the supplied color comparison table. Test strips assume a particular color on contact with the sample solution.

  • Instant pH readings
  • Accurate for a wide range of routine pH testing
  • Inexpensive
  • Convenient and portable for field use. These pH indicators have been impregnated with a mixture of several indicators
TRUSTED_SUSTAINABILITY

Specifications

pH Range 2 to 9
Format Strip
Includes Color comparison chart
Quantity 100 Pack
Type Indicator Paper
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.