Learn More
Cytiva Whatman™ pH Indicators Integral Comparison Strips
Used for rapid determination of pH values
Brand:  Cytiva 2628-990
 View more versions of this product
Product Code. 10614282
  • £30.10 / Pack of 200
Estimated Shipment: 09-10-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
Used for rapid determination of pH values
  • Instant pH readings
  • Accurate for wide range of routine pH testing
  • Convenient and portable for field use
TRUSTED_SUSTAINABILITY
Specifications
Specifications
5.2 to 6.8
Routine pH testing
pH Indicator Test Strip, Integral Comparison Strip
Strip
pH indicator and test papers
100 x 11 mm