Learn More
Cytiva Whatman™ pH Indicators Integral Comparison Strips
Used for rapid determination of pH values
Brand: Cytiva 2629-990
30.01 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
Used for rapid determination of pH values
- Instant pH readings
- Accurate for wide range of routine pH testing
- Convenient and portable for field use
Specifications
6 to 8.1 | |
Routine pH testing | |
pH Indicator Test Strip, Integral Comparison |
Strip | |
Plastic supports, paper segments on each strip, complementary indicator charts | |
100 x 11 mm |