missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ pH Indicators Integral Comparison Strips

Product Code. 11340694
Change view
Click to view available options
pH Range:
1 to 12
1.8 to 3.8
3.8 to 5.5
5.2 to 6.8
6 to 8.1
8 to 9.7
9.5 to 12
Unit Size:
Pack of 200
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. pH Range unitSize
11340694 8 to 9.7 Pack of 200
10451602 3.8 to 5.5 Pack of 200
11370684 1 to 12 Pack of 200
10114071 1.8 to 3.8 Pack of 200
10614282 5.2 to 6.8 Pack of 200
11330694 6 to 8.1 Pack of 200
11914556 9.5 to 12 Pack of 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 11340694 Supplier Cytiva Supplier No. 2630990
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Used for rapid determination of pH values

  • Instant pH readings
  • Accurate for wide range of routine pH testing
  • Convenient and portable for field use
TRUSTED_SUSTAINABILITY

Specifications

pH Range 8 to 9.7
Format Strip
For Use With (Application) Routine pH testing
Includes Plastic supports, paper segments on each strip, complementary indicator charts
Type pH Indicator Test Strip, Integral Comparison Strip
Dimensions (L x W) 100 x 11 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.