missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ pH Indicators Integral Comparison Strips
Description
Used for rapid determination of pH values
- Instant pH readings
- Accurate for wide range of routine pH testing
- Convenient and portable for field use
Specifications
Specifications
| pH Range | 1.8 to 3.8 |
| Format | Strip |
| For Use With (Application) | Routine pH testing |
| Includes | Plastic supports, paper segments on each strip, complementary indicator charts |
| Type | pH Indicator Test Strip, Integral Comparison Strip |
| Dimensions (L x W) | 100 x 11 mm |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?