Learn More
Cytiva Whatman™ pH Indicator, 1.0 to 11.0 Range, 10 books of 10 strips, pH Indicators and Test Papers
Accurate for a wide range of routine pH testing
Brand: Cytiva 2600-500
101.05 GBP valid until 2024-12-31
Use promo code "23266" to get your promotional price.
Code : 69
Additional Details : Weight : 0.35280kg
Description
Accurate for a wide range of routine pH testing
- Instant pH readings.
- Accurate for a wide range of routine pH testing.
- Inexpensive.
- Convenient and portable for field use.
Specifications
Congo Red | |
Strip | |
1 pH | |
pH Indicator Book |
1 to 11 | |
For the rapid determination of pH values has led to many applications in laboratories and industry | |
20 wide-range (pH 1 to 11) strips graduated in 1 pH units, color-comparison chart inside cover, 20 leaves per book, 10 sets of 10 books per pack |