missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ Nuclepore™ PVP-Free Polycarbonate Hydrophobic Cell Culture Membrane
Description
- Low protein binding and very low extractables ensuring no sample contamination
- High chemical resistance and good thermal stability for a wide range of samples
- Low, consistent ash and low tare weights
- Smooth flat surface for good visibility of particles
Specifications
Specifications
| Material | Polycarbonate |
| Pore Size | 8 μm |
| Flow Rate | High |
| For Use With (Application) | Epifluorescence microscopy, environmental analysis, cell biology |
| Diameter (Metric) | 13 mm |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?