missing translation for 'onlineSavingsMsg'
Learn More

Whatman™ Nuclepore™ Polycarbonate Hydrophilic Membranes

Manufactured from high-quality polycarbonate film and track-etched for high pore size precision. Whatman™ Polycarbonate Hydrophilic Membranes combine high flow rates and chemical and thermal resistance with sharply defined pores, making them suitable for microscopy, cytology, parasitology, and certain air and oceanography analyses.

Brand:  Whatman™ 112803

Code : A0

Additional Details : Weight : 0.34000kg

Product Code. 11398964

  • £298.40 / Pack of 25


Estimated Shipment
06-03-2018
Add to basket

Description

Description

Offers a highly defined pore size cutoff in the sub-micron range, enabling particle deformity measurement and capture of extremely small particles.
  • Track-etched for highly defined pore size and high pore density
  • Treated with PVP to render it hydrophilic
  • Low extractables, minimizing sample contamination risk
  • High chemical resistance and good thermal stability for a wide range of sample types
  • Low, consistent ash and low tare weights
  • Smooth flat surface for high particle visibility
Specifications

Specifications

0.05μm
293mm
Hydrophilic
Nuclepore Polycarbonate
25Pack
Track-etched
Videos