missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ Non-sterile ME 25 Mixed Cellulose Ester Membranes

Offers a larger adsorption of product with a high degree of internal surface area. Whatman™ Non-sterile ME25 Mixed Cellulose Membranes are available plain for automatic colony counting applications, as well as gridded to assist in manual counting procedures.

Brand:  Cytiva 10409714

Code : A0

Additional Details : Weight : 0.01100kg


 View more versions of this product

Product Code. 11325394

  • £134.00 / Pack of 100
Estimated Shipment: 21-06-2024
to see stock.
Add to basket

Description

Description

Offers a larger adsorption of product with a high degree of internal surface area. Whatman™ Non-sterile ME25 Mixed Cellulose Membranes are available plain for automatic colony counting applications, as well as gridded to assist in manual counting procedures.

Composed of cellulose acetate and cellulose nitrate characterized by a smooth and uniform surface. Color contrast provided by the filter surface facilitates particle detection and minimizes eye fatigue.
  • Gridded surface (3.1mm intervals) make filters ideal for colony counting after incubation
  • Special nontoxic ink used is completely free from bacterial growth inhibitors
  • Manufactured under strictly controlled cleanroom conditions
  • Pore size: 0.45μm
Specifications

Specifications

Autoclavable
Mixed Cellulose Ester
0.45 μm
White
100 Pack
Grid
130°C
Membrane
2 in.
Black
ME 25
Non-sterile
Medium
3.1mm
266°F
145 μm
12.5 sec./100mL/12.5cm2
50 mm
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • ME5/31, 0.45µm 50mm

Signal Word
  • Warning

Hazard Category
  • Flammable solid Category 2

Hazard Statement
  • H228-Flammable solid.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

Supplemental information
  • MIXTURE LIST-Contains : nitrocellulose

SDS
Documents

Documents

Certificates
Special Offers

Special Offers