Learn More
Cytiva Whatman™ Minifold I Spot-Blot System, complete 96-well (acrylic)
Minifold I Spot-Blot system, complete 96 well (acrylic)
Brand: Cytiva 10447850
Code : A0
Additional Details : Weight : 1.90000kg
Description
- Easy-to-assemble system.
- Available in three interchangeable configurations for spot-, dot-, and slot-blots.
- Accepts all types of transfer membranes: Allows for choice of membrane with the highest binding capacity.
Specifications
Minifold I Spot-Blot System | |
Acrylic 96-well manifold apparatus, 5 x Protran BA85 sheets, 5 x 3mm Chr sheets | |
Acrylic | |
200 μL/well |