missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ ME 25 Plain Mixed Cellulose Ester Membranes

Product Code. 11395304
Change view
Click to view available options
Diameter (Metric):
25 mm
50 mm
90 mm
100 mm
110 mm
142 mm
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 100
Pack of 25
Pack of 50
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Sterility unitSize
11395304 100 mm Non-sterile Pack of 50
11355304 25 mm Non-sterile Pack of 100
11315314 142 mm Non-sterile Pack of 25
11375304 50 mm Non-sterile Pack of 100
11305314 110 mm Non-sterile Pack of 50
11345314 50 mm Sterile Pack of 100
11385304 90 mm Non-sterile Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 11395304 Supplier Cytiva Supplier No. 10401621

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Facilitates the counting process and minimizes eye fatigue with color contrast between the surface and the colonies. Whatman™ ME 25 Plain Mixed Cellulose Ester Membranes are characterized by a smooth and more uniform surface.

  • Excellent contrast for easier particle detection.
  • Offers a high degree of internal surface area for greater adsorption of product.
  • Higher dirt loading capacity.
  • Biologically inert with good thermal stability.
  • No surfactants to contaminate samples.
  • Uniform microporous structure of membrane gives high flow rates.
TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Autoclavable
Bubble Point 2.8 bar
Material Cellulose Acetate, Cellulose Nitrate
Pore Size 0.45 μm
Sterility Non-sterile
Flow Rate 12.5 sec./100 mL/12.5 cm2
Includes Interleaving paper
Membrane Type ME25
Quantity 50/Pk.
Surface Type Plain
Thickness (Metric) 145 μm
Diameter (Metric) 100 mm
Show More Show Less
compliance-icons
Product Identifier
  • ME25, 0.45µm 100mm
Signal Word
  • Warning
Hazard Category
  • Flammable solid Category 2
Hazard Statement
  • H228-Flammable solid.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
  • MIXTURE LIST-Contain: nitrocellulose
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.