missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ Litmus Blue Test Paper

Product Code. 11334974
Click to view available options
Format:
Reel
Strip
Unit Size:
Each
Pack of 100

Product Code. 11334974

Brand: Cytiva 10360300

Only null left
Add to basket
Edge
Add to basket

Impregnated with a mixture of pH indicators, acid-alkali litmus papers assume a color specific to sample pH upon contact with a sample. Whatman™ Litmus Blue Test Paper provides rapid determination of pH values and has a variety of applications in laboratories and industry.

Mixed-indicator tests designed for fast, accurate, and reliable pH solution testing.
  • Instant pH readings
  • Accurate for a wide range of routine pH testing
  • Convenient and portable for field use
  • Color change in pH solutions ranging from approximately 5 to 8
  • Economical
TRUSTED_SUSTAINABILITY

Specifications

Color Indicator Blue
pH Range 5 to 8
Format Strip
Quantity 100 Pack
Type Test Paper
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.