missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman Grade GF/C Glass Microfiber Filters, Binder Free, Ready to Use

Product Code. 15863704
Change view
Click to view available options
Diameter (Metric):
4.7 cm
7.0 cm
9.0 cm
Type:
Filter, pre-rinsed and dried
RTU Filter, pre-weighed
Unit Size:
Pack of 100
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Type unitSize
15863704 7.0 cm Filter, pre-rinsed and dried Pack of 100
15803714 4.7 cm RTU Filter, pre-weighed Pack of 100
15813714 7.0 cm RTU Filter, pre-weighed Pack of 100
15823714 9.0 cm RTU Filter, pre-weighed Pack of 100
15853704 4.7 cm Filter, pre-rinsed and dried Pack of 100
15873704 9.0 cm Filter, pre-rinsed and dried Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 15863704 Supplier Cytiva Supplier No. 2822070

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Grade GF/C glass filters offer fine particle retention, good flow rate and loading capacity, and support total suspended solids analysis.

  • Good flow rate and loading capacity
  • Fine nominal particle retention of 1.2μm
  • Binder free
  • Applications include total suspended solids analysis, industrial waste analysis and potable water
  • GF/C RTU pre-treats traditional GF/C filter with washing, drying, cooling (in a desiccator), and weighing steps aligned to EN 872
  • Certified weight loss of less than 0.017mg/cm2 after preparation
  • Each RTU filter is placed in an individually barcoded aluminum pan that notes the filter weight
TRUSTED_SUSTAINABILITY

Specifications

Airflow Rate 6.7s/100mL/in2
Description GE’s ready-to-use Whatman filter paper Grade GF/C glass filters offer a good flow rate and loading capacity that support total suspended solids analysis with a fine nominal particle retention of 1.2 μm.
Material Borosilicate Glass
Particle Retention 1.2 μm
Pore Size 1.2 μm
For Use With (Application) Widely used for cell harvesting, liquid scintillation counting and binding assays where more loading capacity is required.Highly suitable for use in applications involving air filtration and for gravimetric analysis of volatile materials where ignition is
Grade Grade GF/C
Quantity 100 Pieces
Temperature (Metric) Max 550°C
Thickness (Metric) 260 μm
Type Filter, pre-rinsed and dried
Waterflow Rate 105 mL/min
Diameter (Metric) 7.0 cm
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • FILTER GLASS CIRCLE
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: glass, oxide, chemicals
  • UNKNOWN ACUTE-100 per cent of the mixture consists of component(s) of unknown acute toxicity.
  • UNKNOWN AQUATIC-Contains 100% of components with unknown hazards to the aquatic environment.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.