missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ Grade 93 Wet Strength Qualitative Filter Papers - Circles

Wet-strengthened qualitative circles

Brand:  Cytiva 1093-6215

119.31 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 11302885

  • £144.00 / Pack of 1000
Estimated Shipment: 04-10-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Wet-strengthened qualitative circles

  • Flowrate and retention fall between those for Whatman Grades 1 and 4
  • Max particle retention in liquid of 10 micron at 98% efficiency
  • Not suitable for Kjeldahl estimations as stabilising resins used contain nitrogen

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Grade 93 Qualitative Filter Paper Wet-Strengthened, circle, 150 mm (10 bags of 100 circles)
Cellulose
80 sec.
Grade 93
145 μm
5.9 in.
Disc
10 μm
Intermediate in speed and retention between Grades 1 and 4.
1000 Pieces (10 bags of 100 circles)
Ø 150 mm
150 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers