missing translation for 'onlineSavingsMsg'
Learn More

GE Healthcare Whatman Grade 603 standard cellulose extraction thimble, 28 × 60mm, thickness 1.5mm

Brand:  GE Healthcare Whatman 10350225

Grade 603 standard cellulose extraction thimble, 28 × 60mm, thickness 1.5mm

Code : 69

Additional Details : Weight : 0.13000kg

Product Code. 11313075

  • £58.99 / Pack of 25


Estimated Shipment
08-05-2018
Add to basket