missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ Grade 287 1/2 Folded Filter Paper

Folded filter paper saves time, increases loading capacity

Brand:  Cytiva 10310247

Code : A0

Additional Details : Weight : 0.23000kg

 View more versions of this product

Product Code. 11338894

  • £65.30 / Pack of 50
Estimated Shipment: 21-05-2024
to see stock.
Add to basket

Description

Description

  • Kieselguhr paper with medium to slow flow
  • For separation of very fine semi-colloidal turbidity
  • For clarifying milk serum, starch or sugar-coated solutions, soil suspensions

Specifications

Specifications

Grade 287 ½ Qualitative Filter Paper Folded (Prepleated), 185 mm
For clarifying milk serum, starch solutions, soil suspensions or sugar-containing solutions prior to polarimetry or refractometry
185 mm
Approx. 330 s
Grade 287 ½
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers