missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ Grade 287 1/2 Folded Filter Paper
Folded filter paper saves time, increases loading capacity
Brand: Cytiva 10310244
Code : A0
Additional Details : Weight : 0.12000kg
Description
- Kieselguhr paper with medium to slow flow
- For separation of very fine semi-colloidal turbidity
- For clarifying milk serum, starch or sugar-coated solutions, soil suspensions
Specifications
Grade 287 ½ Qualitative Filter Paper Folded (Prepleated), 125 mm | |
For clarifying milk serum, starch solutions, soil suspensions or sugar-containing solutions prior to polarimetry or refractometry | |
125 mm |
Approx. 330 s | |
Grade 287 ½ |