missing translation for 'onlineSavingsMsg'
Learn More

Cytiva (Formerly GE Healthcare Life Sciences) Whatman™ Glass Fiber Filters, Grade GF/D

Product Code. 16676095 Shop All Cytiva Products
Change view
Click to view available options
Diameter (Metric):
15 mm
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) unitSize
16676095 15 mm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16676095 Supplier Cytiva (Formerly GE Healthcare Life Sciences) Supplier No. 182310157

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For general-purpose filtrations, solvent filtrations, and as a prefilter to extend the life of membrane filters.

TRUSTED_SUSTAINABILITY

Specifications

Material Borosilicate Glass
For Use With (Application) General-Purpose Filtrations, Solvent Filtrations, and as a Prefilter
Grade Grade GF/D
Quantity 100/Pk.
Temperature (Metric) Max 500°C
Diameter (Metric) 15 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.