missing translation for 'onlineSavingsMsg'
Learn More

QIAGEN QIAcard FTA DMPK

Product Code. 15199798
Click to view available options
Product Type:
QIAcard FTA DMPK-A
QIAcard FTA DMPK-B
QIAcard FTA DMPK-C
Formulation:
Type A
Type B
Type C
Unit Size:
Each
Pack of 100
This item is not returnable. View return policy

Product Code. 15199798

Brand: QIAGEN WB129241

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For collection, stabilization, processing, transport and archiving of biological samples. 4 sample areas per card.

QIAcard FTA DMPK Cards for Drug Metabolism Pharmacokinetic enable microvolume sampling with only 10 to 20 μL per sample and are designed for use with blood samples. The cards can also be used with colorless samples such as plasma, cerebrospinal fluid and urine.

QIAcard FTA DMPK-A and QIAcard FTA DMPK-B contains impregnated chemicals enabling protein denaturation to inactivate endogenous enzymes. Cell lysis releases endogenous cellular materials onto card. Stabilization of DNA allows resampling of blood spot for pharmacogenomics.Impregnated chemicals may interfere with mass spectrometry detection (e.g. ion suppression).

QIAcard FTA DMPK-C contain pure cellulose. No impregnated chemicals to interfere with analysis. Proteins are not denatured so cards may be better suited for protein based biomolecules and antibody studies.

Analysis:

Analysis primarily with HPLC or UHPLC followed by MS/MS detection, direct desorption MS techniques such as DESI. Methanol or aqueous/organic mixtures have been shown to work well for small molecules and aqueous buffers are recommended for proteins.

Select a good solvent for the analyte while maintaining compatibility with the HPLC conditions to be used. The choice of DMPK card depends on many factors such as the analyte chemical structure, extraction solvent and analysis workflow.

TRUSTED_SUSTAINABILITY

Specifications

Format Card
Formulation Type A
For Use With (Application) For protein-based biomolecules, on-substrate cleanup has potential to convey greater analyte stability, requires fewer experimental animals
Includes 100 cards
Key Functions Contains impregnated chemicals enabling protein denaturation to inactivate endogenous enzymes. Cell lysis releases endogenous cellular materials onto card. Stabilization of DNA allows resampling of blood spot for pharmacogenomics.
Product Type QIAcard FTA DMPK-A
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.