Learn More
QIAGEN Indicating Desiccant Pack
Product Code. 10111442
Change view
Click to view available options
Quantity:
1 g
Unit Size:
Pack of 1000
Product Code. Quantity unitSize
10111442 1 g Pack of 1000
1 options
Product Code. 10111442 Supplier QIAGEN Supplier No. WB100003
For contamination-free collection, storage and transport of DNA samples.
  • Desiccant packets (1 g) with indicator to ensure that FTA Cards remain dry during transport or storage.
  • A color change from blue to pink indicates absorption of moisture.
Specifications
For Use With (Application) Collection, storage, processing and shipping FTA™ and FTA™ elute cards
Product Type Dessicant Packet
Quantity 1 g