Learn More
QIAGEN EasiCollect Plus
Description
- 50 buccal sample collection devices:
- Each EasiCollect Plus comprise of an integral foam applicator for direct sample transfer to an EasiCollect Card.
- The collection device is closable and designed for consistent and reliable sample transfer and secure transport.
Specifications
Specifications
| Description | EasiCollect + Buccal Sample Collection Device |
| Device Holds | One Sample area and one cleaning punch area per card |
| For Use With (Application) | Forensics and Human Identification |