missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ Binder-Free Glass Microfiber Filters, Grade GF/A

Product Code. 10770802
Change view
Click to view available options
Diameter (Metric):
13 mm
21 mm
24 mm
25 mm
37 mm
42.5 mm
47 mm
50 mm
55 mm
60 mm
70 mm
81 mm
90 mm
110 mm
125 mm
150 mm
240 mm
Unit Size:
Pack of 100
17 product options available for selection
Product selection table with 17 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) unitSize
10770802 25 mm Pack of 100
10102394 13 mm Pack of 100
10644561 21 mm Pack of 100
11330554 24 mm Pack of 100
10251791 37 mm Pack of 100
10577901 42.5 mm Pack of 100
11969144 47 mm Pack of 100
10101391 50 mm Pack of 100
11390554 55 mm Pack of 100
10317871 60 mm Pack of 100
11320564 70 mm Pack of 100
11350354 81 mm Pack of 100
11330564 90 mm Pack of 100
10343731 110 mm Pack of 100
10529441 125 mm Pack of 100
11360564 150 mm Pack of 100
11330354 240 mm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 17 options available.
17 options
This item is not returnable. View return policy
Product Code. 10770802 Supplier Cytiva Supplier No. 1820025

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Combines high loading capacity with fine particle retention. Whatman™ Binder-Free Glass Microfiber Filters, Grade GF/A are ideal for gravimetric determination of airborne particulates, stack sampling and absorption methods of air pollution monitoring.

Whatman glass microfiber filters without binder feature low resistance to fluid flow and high particle-loading capacity. These filters can be used at temperatures up to 500°C. The fine capillary structure of glass fiber filters can absorb large quantities of water, enabling spot tests and liquid scintillation counting methods.

Glass microfiber filters are mainly used in applications such as air or water monitoring and can be used as pre-filters for membranes.

  • High flow rate
  • Good loading capacity
  • Water pollution effluent monitoring
  • Filtering water, algae, bacterial cultures
  • Foodstuff analyses
  • Protein filtration
  • Radioimmunoassay of weak β-emitters
TRUSTED_SUSTAINABILITY

Specifications

Description GE’s Whatman Grade GF/A glass microfiber filters feature fine retention. GF/A filters are suitable for water quality parameter measurement and air filtration applications.
Format Disc
Material Borosilicate Glass
Particle Retention 1.6 μm
Flow Rate 13 sec./100mL
For Use With (Application) Used for high-efficiency general purpose laboratory filtration, including water pollution monitoring of effluents, for filtration of water, foodstuff analyses, protein filtration, and radioimmunoassay of weak β emitters.
Grade Grade GF/A
Quantity 100 Pieces
Shape Circle
Temperature (English) Max. 932°F
Temperature (Metric) Max 500°C
Thickness (English) 0.01 in.
Thickness (Metric) 260 μm
Type Fine Retention Filter
Waterflow Rate 143 mL/min
Diameter (English) 0.98 in.
Diameter (Metric) 25 mm
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Grade GF/A Fine Retention Filter, 25 mm circle (100)pcs
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: glass oxide
  • UNKNOWN ACUTE-100 per cent of the mixture consists of component(s) of unknown acute toxicity.
  • UNKNOWN AQUATIC-Contains 100% of components with unknown hazards to the aquatic environment.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.