missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ Autovial Syringeless Filters::Autovial 12::Without Prefilter
For removing particulates from potentially hazardous samples
£216.00
Specifications
Pore Size | 0.45 μm |
---|---|
Filtration Area | 3 cm2 |
Hold up Volume (After Air Purge) | 140 μL |
Housing Material | Polypropylene (PP) |
Outlet Style | Male slip luer |
Product Code | Brand | Material | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Material | Price | Quantity & Availability | |||||
11984436
|
Cytiva
AV125NPUAQU |
PVDF |
£216.00
Pack of 50 |
Estimated Shipment: 23-07-2024
Log in to see stock. |
|||||
11964436
|
Cytiva
AV125NPUPSU |
PES |
£216.00
Pack of 50 |
Estimated Shipment: 08-07-2024
Log in to see stock. |
|||||
Description
Support stand protects slip-Luer tip, recessed to reduce chance of contamination. Pour or pipet sample into filter barrel, insert plunger and compress to filter into autosampler vial or fit a needle to its outlet for direct injection.
- Handle samples up to 12mL in volume
- No assembly required
- Replace syringe-coupled devices with single filter units for one-time use
- Preassembled units consist of a filter barrel and a plunger
- Built-in air purge minimizes holdup
- Integrated construction eliminates filter pop-off
- Available in PES and PVDF
Polyvinylidene Fluoride (PVDF)
Sample typr: Low protein binding. For aqueous samples, samples ≤60% organic solvent
Polyethersulfone (PES)
For aqueous samples, low protein binding
Specifications
0.45 μm | |
140 μL | |
Male slip luer | |
Syringeless Filter |
3 cm2 | |
Polypropylene (PP) | |
No | |
Up to 12 mL |