Learn More
Cytiva Whatman™ AS 600 Series Multiple Vacuum Filtration Apparatus
AS610/3 Multiple Vacuum Filtration Apparatus, six-place vacuum filtration system, stainless steel 500 ml, 47/50 mm, support screen, with rapid closure clamp
Brand:  Cytiva 10444850
Code : A0
Additional Details : Weight : 7.50000kg
 View more versions of this product
Product Code. 11373165
  • £5820.00 / Each
Estimated Shipment: 19-06-2024
to see stock.
Add to basket
Description
Description
AS 600 Series
The apparatus can be autoclaved and sterilized by dry heat at up to 180°C. Suitable only for vacuum operation. If flushing tubes are used, do not exceed 1.3 bar (300 mbar over-pressure).
Specifications
Specifications
Autoclavable
Stainless Steel
AS600/5 Series
Vacuum filtration system
Stainless Steel support screen filter, 6 place manifold, rapid closure clamp
100 mL