Learn More
Cytiva Whatman™ AS 300 Series Multiple Vacuum Filtration Apparatus
Product Code. 11333165
Change view
Click to view available options
Volume (Metric):
100 mL
500 mL
Includes:
Stainless Steel support screen filter, 3 place manifold
Stainless Steel support screen filter, 3 place manifold, rapid closure clamp
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Includes unitSize
11333165 500 mL Stainless Steel support screen filter, 3 place manifold, rapid closure clamp Each
11323165 500 mL Stainless Steel support screen filter, 3 place manifold Each
11399274 100 mL Stainless Steel support screen filter, 3 place manifold, rapid closure clamp Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11333165 Supplier Cytiva Supplier No. 10445835
Only null left
Add to basket
Add to basket
AS300/3 Multiple Vacuum Filtration Apparatus, three-place vacuum filtration system, stainless steel 500 ml, 47/50 mm, support screen
The apparatus can be autoclaved and sterilized by dry heat at up to 180°C. Suitable only for vacuum operation. If flushing tubes are used, do not exceed 1.3 bar (300 mbar over-pressure).
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
Material Stainless Steel
Includes Stainless Steel support screen filter, 3 place manifold, rapid closure clamp
Series AS310/3
Type Vacuum Filtration System
Volume (Metric) 500 mL