Learn More
Cytiva Whatman™ Antibiotic Assay Paper
Product Code. 11363155
Change view
Click to view available options
Diameter (Metric) Outside:
9 mm
Unit Size:
Pack of 1000
Product Code. Diameter (Metric) Outside unitSize
11363155 9 mm Pack of 1000
1 options
Produktkod. 11363155 Leverantör Cytiva Leverantörsnummer 2017009
Bara null kvar
Lägg i varukorgen
Lägg i varukorgen
Denna artikel kan inte returneras. Se returpolicy
For determining the type of causal agent of infectious diseases and for checking their sensitivity to antibiotics and chemotherapeutic agents in vitro by means of the inhibition zone determination method.
Antibiotic assay discs
The antibiogram allows rational and selective chemotherapy. The test discs can be coated with chemotherapeutic agents, placed on the innoculated nutrient agar and incubated. The size of the inhibition zone is a measure for the effectiveness of the substances.
TRUSTED_SUSTAINABILITY
Specifikationer
Product Type Antibiotic Assay Paper
For Use With (Application) For research use, in determining the type of causal agent of infectious diseases and for checking their sensitivity to antibiotics and chemotherapeutic agents in vitro by means of the inhibition zone determination method
Diameter (Metric) 9 mm
Diameter (Metric) Outside 9 mm
Produkttitel
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.