Learn More
Cytiva Whatman™ Anotop™ 10mm Syringe Filters
Removes virtually 100% of particulates larger than pore size. Whatman™ Anatop™ 10mm Syringe Filters include a highly defined capillary pore structure and narrow pore-size distribution, enabling a high level of consistency in particle retention.
£486.00 - £916.00
Specifications
Disposable | Single-use |
---|---|
Material (Membrane) | Anopore Inorganic Membrane (alumina matrix) |
Sterility | Non-sterile |
Filtration Area | 0.78 cm2 |
For Use With (Application) | HPLC aqueous/organic sample preparation; cold sterilization of growth media; solvent filtration for spectroanalysis; phage and virus filtration; liposome extrusion; removal of high molecular weight proteins or polymers; colloid removal from deionized water |
Product Code | Brand | Pore Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | Price | Quantity & Availability | |||||
11394704
|
Cytiva
6809-1012 |
0.1 μm |
£486.00
Pack of 50 |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||||
11304714
|
Cytiva
6809-1022 |
0.2 μm |
£515.00
Pack of 50 |
Estimated Shipment: 20-05-2024
Log in to see stock. |
|||||
11384704
|
Cytiva
6809-1002 |
0.02 μm |
£916.00
Pack of 50 |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
Description
Removes virtually 100% of particulates larger than pore size. Whatman™ Anatop™ 10mm Syringe Filters include a highly defined capillary pore structure and narrow pore-size distribution, enabling a high level of consistency in particle retention.
Compatible with aqueous suspensions and a wide range of organic solvents. Handle volumes up to 5mL (10mm diameter filter) or 100mL (25mm diameter filter).
- Nondeformable, honeycombed Anopore™ membrane
- Inherently hydrophilic membrane is free of adhesives and surfactants
- Low protein binding, low fluid retention for maximum sample recovery
- Unidirectional flow
Order Info
Hexagonal, thermal-welded polypropylene housing is free of adhesives and binders.
Specifications
Single-use | |
Non-sterile | |
HPLC aqueous/organic sample preparation; cold sterilization of growth media; solvent filtration for spectroanalysis; phage and virus filtration; liposome extrusion; removal of high molecular weight proteins or polymers; colloid removal from deionized water | |
Polypropylene | |
Male Luer Slip | |
50 Pack | |
Syringe Filter | |
10 mm |
Anopore Inorganic Membrane (alumina matrix) | |
0.78 cm2 | |
<20 μL | |
Female Luer Lock | |
6.9bar (100psi) | |
45°C | |
Up to 10 mL |