missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ Acid-Treated Low-Metal TCLP Filters

Binder free filters

Brand:  Cytiva 1810-090

Code : A0

Additional Details : Weight : 0.05000kg

 View more versions of this product

Product Code. 11333135

  • £99.50 / Pack of 50
Estimated Shipment: 26-09-2024
to see stock.
Add to basket

Description

Description

Binder free filters

The Whatman TCLP Filter is manufactured using a binder free borosilicate glass microfiber with a particle retention rating of 0.6μm to 0.8μm

  • Resistant to acids and bases
  • Available in six diameters, including sizes to fit Büchner funnels and zero-headspace extraction units
  • Pretreatment with acid eliminates need to acid-wash filters in your lab, for reduced analysis time and reagent costs

Compliance

Conform to requirements of U.S. EPA Method 1311 Toxicity Characteristic Leaching Procedure (TCLP), including 0.6 to 0.8μm particle retention requirement.

Specifications

Specifications

GE’s Whatman TCLP test filters are acid-treated with low metal content. They support TCLP testing by providing accurate information about TCLP limits to ensure the best possible disposal plan can be put into action.
Borosilicate Glass
Required for volatile samples and use with a zero headspace extractor.
Circle
TCLP Limits Test Filter
3.5 in.
Disc
0.7 μm
50 pieces
420 μm
60 mL/min
90 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers