missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ Acid-Treated Low-Metal TCLP Filters

Binder free filters

Brand:  Cytiva 1810-047

Code : A0

Additional Details : Weight : 0.38000kg

 View more versions of this product

Product Code. 11323135

  • £125.00 / Pack of 100
Estimated Shipment: 21-05-2024
to see stock.
Add to basket

Description

Description

Binder free filters

The Whatman TCLP Filter is manufactured using a binder free borosilicate glass microfiber with a particle retention rating of 0.6μm to 0.8μm

  • Resistant to acids and bases
  • Available in six diameters, including sizes to fit Büchner funnels and zero-headspace extraction units
  • Pretreatment with acid eliminates need to acid-wash filters in your lab, for reduced analysis time and reagent costs

Compliance

Conform to requirements of U.S. EPA Method 1311 Toxicity Characteristic Leaching Procedure (TCLP), including 0.6 to 0.8μm particle retention requirement.

Specifications

Specifications

GE’s Whatman TCLP test filters are acid-treated with low metal content. They support TCLP testing by providing accurate information about TCLP limits to ensure the best possible disposal plan can be put into action.
Borosilicate Glass
Determine the leaching potential in a landfill for hazardous organic and inorganic contaminants that could potentially migrate into groundwater, threatening drinking water sources
Circle
TCLP Limits Test Filter
1.8 in.
Disc
0.7 μm
100 pieces
420 μm
60 mL/min
47 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers