missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ 0.6 μm Pore Size (ME 26) Mixed Cellulose Ester Membranes, Plain
Whatman™ (ME 26) Mixed Cellulose Ester Membranes are particularly effective in applications requiring higher flow rates and larger volume filtration, including clarification or sterilization of aqueous solutions, particulate analysis and removal, air monitoring, and microbial analysis.
£103.00 - £138.00
Specifications
Autoclavable | Autoclavable |
---|---|
Bubble Point | 3.7 bar |
Disposable | Single-use |
Filter Type | Plain |
Material | MCE |
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
11345014
|
Cytiva
10401514 |
50 mm |
£138.00
Pack of 100 |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||||
11315014
|
Cytiva
10401506 |
25 mm |
£103.00
Pack of 100 |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||||
Description
- Filter Type: Plain
- Material: Mixed Cellulose Ester
- Material (Membrane): ME 26
- Pore Size: 0.6 μm
- Autoclavable: Yes
- Sterility: Non-sterile
- Flow Rate: 4.8 sec./100 mL/12.5 cm² (at Δp = 0.9 bar)
- Gas Flow Rate: 21 sec./100 mL (at Δp = 3 mbar)
- Bubble Point: 3.7 bar
- For Use With (Application): Filtration, cytology, air monitoring, aqueous HPLC samples, biological assays
Specifications
Autoclavable | |
Single-use | |
MCE | |
0.6 μm | |
4.8 sec./100 mL/12.5cm² (at Δp = 0.9 bar) | |
21 sec./100 mL (at Δp = 3 mbar) |
3.7 bar | |
Plain | |
ME 26 | |
Non-sterile | |
Filtration, cytology, air monitoring, aqueous HPLC samples, biological assays | |
Plain |