missing translation for 'onlineSavingsMsg'
Learn More

WH-4-023, MedChemExpress

Product Code. 16467797
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16467797 5 mg 5mg
16465078 10 mg 10mg
16431317 50 mg 50mg
16415816 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16467797 Supplier Medchem Express Supplier No. HY12299

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress WH-4-023 is a potent and selective dual Lck/Src inhibitor with IC50 of 2 nM/6 nM for Lck and Src kinase respectively; little inhibition on p38α and KDR.

WH-4-023 is a potent and selective dual Lck/Src inhibitor with IC50 of 2 nM/6 nM for Lck and Src kinase respectively

Specifications

Chemical Name or Material WH-4-023
CAS 837422-57-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C32H36N6O4
Quantity 5 mg
Synonym Dual LCK/SRC inhibitor
Solubility Information DMSO : 25 mg/mL (43.96 mM; Need ultrasonic)
SMILES O=C(OC1=C(C)C=CC=C1C)N(C2=CC=C(OC)C=C2OC)C3=NC(NC4=CC=C(N5CCN(C)CC5)C=C4)=NC=C3
Molecular Weight (g/mol) 568.67
Formula Weight 568.67
Percent Purity 99.35%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.