missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ WFDC8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583625
This item is not returnable.
View return policy
Description
Immunogen sequence: KHKPGLCPKE RLTCTTELPD SCNTDFDCKE YQKCCFFACQ KKCMDPFQEP CMLPVRHGNC NHEAQRWHFD FKNYRCTPFK YRGCEGNA Highest antigen sequence indentity to the following orthologs: Rat - 48%, Mouse - 55%.
This gene encodes a member of the WAP-type four-disulfide core (WFDC) domain family. The WFDC domain, or WAP signature motif, contains eight cysteines forming four disulfide bonds at the core of the protein, and functions as a protease inhibitor. The encoded protein contains a Kunitz-inhibitor domain, in addition to three WFDC domains. Most WFDC genes are localized to chromosome 20q12-q13 in two clusters: centromeric and telomeric. This gene belongs to the telomeric cluster. Two alternatively spliced transcript variants have been found for this gene, and they encode the same protein. [provided by RefSeq, Jul 2008].
Specifications
| WFDC8 | |
| Polyclonal | |
| Unconjugated | |
| WFDC8 | |
| C20orf170; dJ461P17.1; epididymis secretory protein Li 292; HEL-S-292; protease inhibitor WAP8; putative protease inhibitor WAP8; testicular secretory protein Li 68; WAP four-disulfide core domain 8; WAP four-disulfide core domain protein 8; WAP motif protein 1; WAP8; WFDC8 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 90199 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8IUA0 | |
| WFDC8 | |
| Recombinant protein corresponding to Human WFDC8. Recombinant protein control fragment (Product #RP-99446). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction