missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WFDC5 Antibody (M47-P2D5), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50363IR
This item is not returnable.
View return policy
Description
WFDC5 Monoclonal specifically detects WFDC5 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
| WFDC5 | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| WFDC5 | |
| Ovalbumin-conjugated synthetic peptide TERVHDGRPG | |
| 0.1 mL | |
| Cancer, Cellular Signaling | |
| 149708 | |
| Store at 4C in the dark. | |
| IgG1 κ |
| Western Blot, ELISA, Immunohistochemistry | |
| M47-P2D5 | |
| Western Blot, ELISA, Immunohistochemistry | |
| dJ211D12.5, PRG5protease inhibitor WAP1, Putative protease inhibitor WAP1, WAP four-disulfide core domain 5, WAP four-disulfide core domain protein 5, WAP1p53-responsive gene 5 protein | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction