missing translation for 'onlineSavingsMsg'
Learn More

WFDC5 Antibody (M47-P2D5) - BSA Free, Novus Biologicals™

Product Code. 18633939 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
Unit Size:
0.03mL
0.1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18633939 0.025 mL 0.03mL
18693859 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18633939 Supplier Novus Biologicals Supplier No. NBP2503630.025ml

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

WFDC5 Monoclonal antibody specifically detects WFDC5 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen WFDC5
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone M47-P2D5
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500
Formulation PBS
Gene Alias dJ211D12.5, PRG5protease inhibitor WAP1, Putative protease inhibitor WAP1, WAP four-disulfide core domain 5, WAP four-disulfide core domain protein 5, WAP1p53-responsive gene 5 protein
Host Species Mouse
Immunogen Ovalbumin-conjugated synthetic peptide TERVHDGRPG
Purification Method Protein A purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Cancer, Cellular Signaling
Primary or Secondary Primary
Gene ID (Entrez) 149708
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.