missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ WFDC2 Recombinant Protein

Human full-length ORF recombinant protein with GST-tag at N-terminal

Brand:  Abnova™ H00010406-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16195022

  • £425.00 / 25µg
Estimated Shipment: 04-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Encoded by WAP four-disulfide core domain 2
  • Molecular weight: 36.08kDa
  • Preparation method: in vitro wheat germ expression system
  • Purification: glutathione sepharose 4 fast flow
  • Storage buffer: 50 mM Tris-HCI, 10 mM reduced glutathione
  • pH=8.0 in elution buffer

Sequence: EKTGVCPELQADQNCTQECVSDSECADNLKCCSAGCATFCSLPNDKEGSCPQVNINFPQLGLCRDQCQVDSQCPGQMKCCRNGCGKVSCVTPNF

Best when used within three months from the date of receipt of this protein.

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

AAH46106.1
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
36.08
8
Glutathione Sepharose 4 Fast Flow
25 ug
EKTGVCPELQADQNCTQECVSDSECADNLKCCSAGCATFCSLPNDKEGSCPQVNINFPQLGLCRDQCQVDSQCPGQMKCCRNGCGKVSCVTPNF
HE4/MGC57529/WAP5/dJ461P17.6
WFDC2
Wheat Germ (in vitro)
GST
Antibody Production, ELISA, Protein Array, Western Blot
10406
WFDC2 (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
WFDC2
Human
Recombinant
Solution
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers