Learn More
Invitrogen™ WFDC2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA580227
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: rat testis tissue, rat brain tissue, rat thymus tissue, mouse testis tissue, mouse brain tissue, mouse thymus tissue, mouse HEPA1-6 whole cell. IHC: mouse lung tissue, mouse small intestine tissue, rat kidney tissue, rat lung tissue, rat small intestine tissue.
This gene encodes a protein that is a member of the WFDC domain family. The WFDC domain, or WAP Signature motif, contains eight cysteines forming four disulfide bonds at the core of the protein, and functions as a protease inhibitor in many family members. This gene is expressed in pulmonary epithelial cells, and was also found to be expressed in some ovarian cancers. The encoded protein is a small secretory protein, which may be involved in sperm maturation.
Specifications
| WFDC2 | |
| Polyclonal | |
| Unconjugated | |
| WFDC2 | |
| 1600023A02Rik; dJ461P17.6; EDDM4; epididymal protein 4; epididymal secretory protein 4; Epididymal secretory protein E4; epididymis-specific, whey-acidic protein type, four-disulfide core; HE4; Major epididymis-specific protein E4; MGC57529; putative protease inhibitor WAP5; RE4; WAP domain containing protein HE4-V4; WAP domain-containing protein HE4; WAP four-disulfide core domain 2; WAP four-disulfide core domain protein 2; WAP5; Wfdc2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 286888, 67701 | |
| -20°C | |
| Lyophilized |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 500 μg/mL | |
| PBS with 4mg trehalose and 0.05mg sodium azide | |
| Q8CHN3, Q9DAU7 | |
| WFDC2 | |
| E. coli-derived rat HE4 recombinant protein (Position: E31-F168). | |
| 100 μg | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.