missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WFDC2, Mouse anti-Human, Clone: 1A23, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00010406-M02.100ug
This item is not returnable.
View return policy
Description
Sequence: EKTGVCPELQADQNCTQECVSDSECADNLKCCSAGCATFCSLPNDKEGSCPQVNINFPQLGLCRDQCQVDSQCPGQMKCCRNGCGKVSCVTPNFSpecifications
| WFDC2 | |
| Monoclonal | |
| Unconjugated | |
| WAP four-disulfide core domain 2 | |
| HE4/MGC57529/WAP5/dJ461P17.6 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA | |
| 1A23 | |
| In 1x PBS, pH 7.4 | |
| BC046106 | |
| WFDC2 | |
| WFDC2 (AAH46106.1, 31 a.a. ∼ 124 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 10406 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction