missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WFDC12 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25101-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the WAP-type four-disulfide core (WFDC) domain family. The WFDC domain, or WAP signature motif, contains eight cysteines forming four disulfide bonds at the core of the protein, and functions as a protease inhibitor. Most WFDC gene members are localized to chromosome 20q12-q13 in two clusters: centromeric and telomeric. This gene belongs to the centromeric cluster.Specifications
| WFDC12 | |
| Polyclonal | |
| Unconjugated | |
| WFDC12 | |
| C20orf122, SWAM2, WAP2, WFDC12, Whey acidic protein 2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 128488 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8WWY7 | |
| WFDC12 | |
| WFDC12 Fusion Protein Ag18795 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction