missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ West Nile Virus Matrix Synthetic Peptide

Synthetic Peptide

Brand:  Invitrogen™ PEP0219

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 15226439

  • £112.00 / 50µg
Estimated Shipment: 11-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

West Nile Virus Matrix Synthetic Peptide, PEP-0219, from Invitrogen.

This peptide corresponds to 15 amino acids near the carboxy terminus of WNV Matrix Protein.
Specifications

Specifications

Virus
West Nile Virus Matrix
-20°C
Liquid
Blocking Assay, Control
0.02% sodium azide
50 μg
PBS
purified
200 μg/mL
Synthetic Peptide
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
No
Peptide
RUO - research use only
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers