missing translation for 'onlineSavingsMsg'
Learn More
Learn More
West Nile Virus Envelope Antibody, mFluor Violet 610 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
£462.00
Specifications
| Antigen | West Nile Virus Envelope |
|---|---|
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | mFluor Violet 610 SE |
| Form | Purified |
Description
West Nile Virus Envelope Polyclonal antibody specifically detects West Nile Virus Envelope in Virus samples. It is validated for ELISASpecifications
| West Nile Virus Envelope | |
| Polyclonal | |
| Purified | |
| RUO | |
| Virus | |
| IgG | |
| Peptide affinity purified |
| ELISA | |
| mFluor Violet 610 SE | |
| Rabbit | |
| Immunology, Virology Bacteria and Parasites | |
| Antibody was raised against a synthetic peptide corresponding to 16 amino acids at the N terminus of the West Nile virus envelope protein. The immunogen is located within amino acids 350 - 400 of West Nile Virus Envelope Protein. Amino Acid Squence: CPTMGEAHNEKRADPA | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title