missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
West Nile Virus Envelope Polyclonal antibody specifically detects West Nile Virus Envelope in Virus samples. It is validated for ELISA
Specifications
Specifications
| Antigen | West Nile Virus Envelope |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | HRP |
| Formulation | PBS |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with purified, recombinant West Nile Virus (lineage 1, strain NY99) Envelope Protein (Domain III) (ABA62343.1; Gln586-Ala705). |
| Purification Method | Antigen and protein A Affinity-purified |
| Quantity | 0.1 mL |
| Regulatory Status | RUO |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?