missing translation for 'onlineSavingsMsg'
Learn More

West Nile Virus Envelope Antibody, DyLight 650, Novus Biologicals™

Product Code. 18355163 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18355163 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18355163 Supplier Novus Biologicals Supplier No. NBP306464C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

West Nile Virus Envelope Polyclonal antibody specifically detects West Nile Virus Envelope in Virus samples. It is validated for ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen West Nile Virus Envelope
Applications ELISA, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant West Nile Virus (lineage 1, strain NY99) Envelope Protein (Domain III) (ABA62343.1; Gln586-Ala705).
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Virology Bacteria and Parasites
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.