Learn More
Thermo Scientific™ Wellcolex™ Color Salmonella Rapid Latex Agglutination Test Kit
Description
Rapidly detect and presumptively identify Salmonella from culture by latex agglutination using Thermo Scientific™ Wellcolex™ Color Salmonella Rapid Latex Agglutination Test Kit. Save up to two days' testing time over traditional screening procedures with this easy-to-use, cost-effective test. With its high positive and negative predictive values (PPV > 98.7%) and results in less than three minutes, laboratories can use this rapid screening test to eliminate Salmonella-negative samples from further testing, reducing the number of samples requiring confirmatory testing.
- Identifies major groups of Salmonella
- Utilizes advanced color technology for clear, easy-to-read species and/or serogroup identification
- Provides accurate, reliable results in less than three minutes
- Quickly eliminates negative samples when used as primary culture screening method
To perform Wellcolex Color Salmonella, a sample from a Selenite F broth culture (18 to 24 hours incubation) or a suspension of bacteria from solid agar is reacted with two test reagents, consisting of a mixture of suspensions of red, blue and green latex particles, each of which is coated with antibody specific for different Salmonella serogroups.
In the presence of homologous antigen one of the colors in the mixture will agglutinate, and the identity of the antigen is indicated by the color of the aggregated particles with a contrasting change in the color of the background. Each combination is easily distinguished from the others and from a negative result in which the particles remain in smooth grey‑brown suspension, and the occasional non‑specific result in which all the particles agglutinate into grey‑brown aggregates against a cleared background.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Description | Wellcolex Colour Salmonella Kit |
| CE Marker | Yes |
| Detectable Analytes | Salmonella |
| Type | Salmonella Test Kit |
| Includes | Latex reagents 1 and 2, red, blue, and green positive controls, disposable mixing sticks, disposable reaction cards, disposable sample dispensers, instructions for use, and reading guide |
| Organism Group | Group A, B, C, D, E and G |
| Quantity | 200 Tests/Kit |
| Target Organism Class | Salmonella |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.